Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Propiolate, MP Biomedicals

Catalog No. ICN215482 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215482 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215482 Supplier MP Biomedicals, Inc Supplier No. 0521548210

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 922-67-8
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.074
InChI Key IMAKHNTVDGLIRY-UHFFFAOYSA-N
Synonym methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
PubChem CID 13536
IUPAC Name methyl prop-2-ynoate
SMILES COC(=O)C#C

Specifications

Quantity 10 g
Formula Weight 84.07
Chemical Name or Material Methyl Propiolate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.