Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Propionate, 99+%

Catalog No. p-167708
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149052500 250 mL Glass bottle
AC149050010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC149052500 Supplier Thermo Scientific Chemicals Supplier No. 149052500
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 554-12-1 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-12-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009306
InChI Key RJUFJBKOKNCXHH-UHFFFAOYSA-N
Synonym methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
PubChem CID 11124
IUPAC Name methyl propanoate
SMILES CCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Melting Point -88°C
Color Colorless
Density 0.9150g/mL
Boiling Point 78°C to 79°C
Flash Point -2°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.377
Linear Formula C2H5CO2CH3
Quantity 250 mL
Beilstein 02, 239
Specific Gravity 0.915
Merck Index 15, 6185
Solubility Information Solubility in water: 5g/L water (20°C). Other solubilities: miscible with alcohol and ether,soluble in most common organic solvents
Formula Weight 88.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Methyl propionate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-060-4
RTECSNumber UF5970000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.