Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl propyl disulfide | 2179-60-4 | 1 ML
SDP

Catalog No. 5000418834
Encompass_Preferred

Methyl propyl disulfide is a volatile sulfur-containing compound primarily found in garlic and onions, recognized for its anticancer properties. It is provided as a liquid with a purity of 98.0%.

  • Volatile sulfur-containing compound
  • Found in garlic and onions
  • Exhibits anticancer effect
  • Liquid appearance
  • Colorless to light yellow
  • Purity of 98.0%
  • Molecular weight of 122.25
  • Chemical formula C4H10S2
  • Stable under recommended storage conditions
  • For research use only

Catalog No. 50-004-18834 Supplier Medchemexpress LLC Supplier No. HYN743610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl propyl disulfide is a volatile sulfur-containing compound primarily found in garlic and onions, recognized for its anticancer properties. It is provided as a liquid with a purity of 98.0%.

  • Volatile sulfur-containing compound
  • Found in garlic and onions
  • Exhibits anticancer effect
  • Liquid appearance
  • Colorless to light yellow
  • Purity of 98.0%
  • Molecular weight of 122.25
  • Chemical formula C4H10S2
  • Stable under recommended storage conditions
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.