Learn More
Medchemexpress LLC Methyl protodioscin | 54522-52-0 | 1063.23 | 1 ML
SDP
Catalog No. 5000416816
Encompass_Preferred
Methyl protodioscin is a multi-target, selective, steroidal diglycoside inhibitor with antitumor activity that induces cell cycle arrest. It exhibits significant antitumor, anti-inflammatory, and anti-restenosis activities, making it suitable for research on various cancers and inflammatory diseases.
  • Induces G2/M cell cycle arrest.
  • Regulates Bcl-2/Bax apoptotic pathway.
  • Inhibits Akt1/c-Myc axis and MAPK/ERK signaling.
  • Reduces cholesterol synthesis by downregulating ADAM15 and inducing FOXO1.
  • Inhibits JNK/c-Jun pathway to reduce inflammatory factors.
  • Shows potent antitumor effects, including inhibiting proliferation, migration, and invasion.
  • Demonstrates anti-inflammatory and anti-restenosis properties.
  • Useful in lung, prostate, and pancreatic cancer research.
  • Applicable in studies of airway inflammation and enteritis.
Catalog No. 50-004-16816 Supplier Medchemexpress LLC Supplier No. HYN086310MM/1ML
Methyl protodioscin is a multi-target, selective, steroidal diglycoside inhibitor with antitumor activity that induces cell cycle arrest. It exhibits significant antitumor, anti-inflammatory, and anti-restenosis activities, making it suitable for research on various cancers and inflammatory diseases.
  • Induces G2/M cell cycle arrest.
  • Regulates Bcl-2/Bax apoptotic pathway.
  • Inhibits Akt1/c-Myc axis and MAPK/ERK signaling.
  • Reduces cholesterol synthesis by downregulating ADAM15 and inducing FOXO1.
  • Inhibits JNK/c-Jun pathway to reduce inflammatory factors.
  • Shows potent antitumor effects, including inhibiting proliferation, migration, and invasion.
  • Demonstrates anti-inflammatory and anti-restenosis properties.
  • Useful in lung, prostate, and pancreatic cancer research.
  • Applicable in studies of airway inflammation and enteritis.