Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl protogracillin | 54522-53-1 | MFCD32197353 | 99.0% | 1079.23 | C52H86O23 | 10 MG
SDP

Catalog No. 5000289791
Encompass_Preferred

Methyl protogracillin is a steroidal saponin natural product isolated from the roots of Dioscorea opposite Thunb. It has reported anticancer activity and is supplied for laboratory research use as a solid with high purity.

  • Steroidal saponin natural product isolated from plant roots.
  • Reported anticancer activity in the literature.
  • High purity suitable for research (99.03%).
  • Supplied as a solid in a 10 mg pack size.
  • Molecular weight 1,079.23; formula C52H86O23.
  • Store at -20°C and protect from light; follow laboratory safety procedures.

Catalog No. 50-002-89791 Supplier Medchemexpress LLC Supplier No. HYN427710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl protogracillin is a steroidal saponin natural product isolated from the roots of Dioscorea opposite Thunb. It has reported anticancer activity and is supplied for laboratory research use as a solid with high purity.

  • Steroidal saponin natural product isolated from plant roots.
  • Reported anticancer activity in the literature.
  • High purity suitable for research (99.03%).
  • Supplied as a solid in a 10 mg pack size.
  • Molecular weight 1,079.23; formula C52H86O23.
  • Store at -20°C and protect from light; follow laboratory safety procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.