Learn More
Medchemexpress LLC HY-N4277 5mg Medchemexpress, Methyl protogracillin CAS:54522-53-1 Purity:>98%
SDP
Catalog No. 501970436
Encompass_Preferred
Medchemexpress, HY-N4277 5mg Methyl protogracillin CAS:54522-53-1 Methyl protogracillin (NSC-698793), isolated from the roots of Dioscorea opposite Thunb, exhibits strong anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0436 Supplier Medchemexpress LLC Supplier No. HYN42775MG
Medchemexpress, HY-N4277 5mg Methyl protogracillin CAS:54522-53-1 Methyl protogracillin (NSC-698793), isolated from the roots of Dioscorea opposite Thunb, exhibits strong anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.