Learn More
Hach Company Methyl Purple Indicator Solution, 500 mL
Catalog No. 501995540 Shop All Hach Company Products
Encompass
1.0 g/L. For alkalinity titrations; pH range 4.8 to 5.4.
Catalog No. 50-199-5540 Supplier Hach Company Supplier No. 2193449
1.0 g/L. For alkalinity titrations; pH range 4.8 to 5.4.