Learn More
Hach Company Methyl Purple Indicator Solution, 100 mL MDB
Catalog No. 501995539 Shop All Hach Company Products
Encompass
1.0 g/L. For alkalinity titrations; pH range 4.8 to 5.4.
Catalog No. 50-199-5539 Supplier Hach Company Supplier No. 2193432
1.0 g/L. For alkalinity titrations; pH range 4.8 to 5.4.