Learn More
Aqua Solutions Methyl Purple Indicator in Methanol (pH 4.8 Purple pH 5.4 Green) (250ML)
SDP
Supplier:  Aqua Solutions 5842250MLP
Encompass_Preferred
Methyl Purple Indicator in Methanol (pH 4.8 Purple pH 5.4 Green) (250ML)
Catalog No. NC0430774
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov