Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Purple Solution pH 4.8(Purple) - pH 5.4(Gray) - pH 5.8(Green), TCI America™
SDP

Catalog No. M0112500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0112500ML 500 mL
M011225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0112500ML Supplier TCI America Supplier No. M0112500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00070843
Synonym Fleisher′s Indicator Solution

Specifications

Color Green
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Methyl Purple Solution pH 4.8(Purple) - pH 5.4(Gray) - pH 5.8(Green)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.