Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl pyrazine-2-carboxylate, 97%

Catalog No. AC443170050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443170050 5 g
AC443170010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443170050 Supplier Thermo Scientific Chemicals Supplier No. 443170050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6164-79-0 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6164-79-0
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
InChI Key TWIIRMSFZNYMQE-UHFFFAOYSA-N
Synonym methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
PubChem CID 72662
IUPAC Name methyl pyrazine-2-carboxylate
SMILES COC(=O)C1=NC=CN=C1

Specifications

Melting Point 59.5°C to 61.5°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 138.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl pyrazine-2-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 228-198-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.