Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Pyrazole-3-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M24441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24441G Supplier TCI America Supplier No. M24441G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15366-34-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00649381
InChI Key ORUCTBNNYKZMSK-UHFFFAOYSA-N
Synonym methyl 1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid methyl ester, methyl pyrazole-3-carboxylate, 2h-pyrazole-3-carboxylic acid methyl ester, 3-methoxycarbonylpyrazole, pyrazole-3-carboxylic acid methyl ester, methyl 2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, methyl ester, pubchem14332, 5-methoxycarbonylpyrazole
PubChem CID 565662
IUPAC Name methyl 1H-pyrazole-5-carboxylate
SMILES COC(=O)C1=CC=NN1

Specifications

Melting Point 146°C
Color White
Quantity 1 g
Formula Weight 126.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Pyrazole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.