Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl pyrimidine-4-carboxylate | 2450-08-0 | 138.12 g/mol | C6H6N2O2 | 1 G
SDP

Catalog No. 5000210744
Encompass_Preferred

Methyl pyrimidine-4-carboxylate is a small-molecule synthetic intermediate used in pharmaceutical and organic synthesis research. It is supplied as a solid intended for research use only.

  • Uses as a synthetic intermediate in pharmaceutical research.
  • CAS number 2450-08-0.
  • Molecular formula C6H6N2O2.
  • Molecular weight 138.12 g/mol.
  • Appearance: solid.
  • Typical storage: room temperature.
  • Supplied as 1 G quantity.

Catalog No. 50-002-10744 Supplier Medchemexpress LLC Supplier No. HYW0433111G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl pyrimidine-4-carboxylate is a small-molecule synthetic intermediate used in pharmaceutical and organic synthesis research. It is supplied as a solid intended for research use only.

  • Uses as a synthetic intermediate in pharmaceutical research.
  • CAS number 2450-08-0.
  • Molecular formula C6H6N2O2.
  • Molecular weight 138.12 g/mol.
  • Appearance: solid.
  • Typical storage: room temperature.
  • Supplied as 1 G quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.