Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl pyrimidine-4-carboxylate | 2450-08-0 | MFCD00234184 | 99.0% | 138.12 g/mol | C6H6N2O2 | 10 G
SDP

Catalog No. 5000210211
Encompass_Preferred

Methyl pyrimidine-4-carboxylate is the methyl ester of pyrimidine-4-carboxylic acid and is used as a synthetic intermediate in pharmaceutical and organic synthesis. It is supplied as a white to light brown solid and is characterized by high purity and standard analytical data for research applications.

  • Molecular formula: C6H6N2O2.
  • Molecular weight: 138.12 g/mol.
  • Cas number: 2450-08-0.
  • Purity: 99.0% (HPLC).
  • Physical state: white to light brown solid.
  • Common package sizes: 1 g, 5 g, 10 g.
  • Intended for research use only.
  • Used as a synthetic intermediate in pharmaceutical and organic synthesis.

Catalog No. 50-002-10211 Supplier Medchemexpress LLC Supplier No. HYW04331110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl pyrimidine-4-carboxylate is the methyl ester of pyrimidine-4-carboxylic acid and is used as a synthetic intermediate in pharmaceutical and organic synthesis. It is supplied as a white to light brown solid and is characterized by high purity and standard analytical data for research applications.

  • Molecular formula: C6H6N2O2.
  • Molecular weight: 138.12 g/mol.
  • Cas number: 2450-08-0.
  • Purity: 99.0% (HPLC).
  • Physical state: white to light brown solid.
  • Common package sizes: 1 g, 5 g, 10 g.
  • Intended for research use only.
  • Used as a synthetic intermediate in pharmaceutical and organic synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.