Learn More
Medchemexpress LLC Methyl pyropheophorbide-a | 6453-67-4 | 99.1% | 548.67 | 25 MG
SDP
Catalog No. 5000350559
Encompass_Preferred
Methyl pyropheophorbide-a is a chlorophyll-a derivative and a potent photosensitizer. It is used in photodynamic therapy (PDT) for cancer, where it exhibits photodynamic activity that induces apoptosis and inhibits tumor growth.
  • Functions as a potent photosensitizer
  • Induces apoptosis and inhibits tumor growth
  • Exhibits dose-dependent photocytotoxicity
  • Inhibits cell cycle progression in cancer cells
  • Demonstrates in vivo tumor growth inhibition
Catalog No. 50-003-50559 Supplier Medchemexpress LLC Supplier No. 25MGHY137473
Methyl pyropheophorbide-a is a chlorophyll-a derivative and a potent photosensitizer. It is used in photodynamic therapy (PDT) for cancer, where it exhibits photodynamic activity that induces apoptosis and inhibits tumor growth.
  • Functions as a potent photosensitizer
  • Induces apoptosis and inhibits tumor growth
  • Exhibits dose-dependent photocytotoxicity
  • Inhibits cell cycle progression in cancer cells
  • Demonstrates in vivo tumor growth inhibition