Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl pyruvate

Catalog No. 371173100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-117-3100G 100 g
37-117-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-117-3100G Supplier Sigma Aldrich Supplier No. 371173100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl pyruvate is a pyruvic ester
  • Surface conditions during pt/silica europt-1 modified by cinchonidine catalyzed by enantioselective hydrogenation of methyl pyruvate to r-(+)-methyl lactate were investigated
  • Enantioselective hydrogenation of methyl pyruvate over finely dispersed polyvinylpyrrolidone-stabilized and the corresponding alumina-supported iridium clusters modified with cinchonidine was studied
  • The molecular orientation and conformation of methyl pyruvate on ni(111) surface has been investigated in the temperature range 105-220k
  • Methyl pyruvate is reported to stimulate insulin secretion in vitro and in vivo.methyl pyruvate may be used to investigate the adsorption and reactivity of methyl pyruvate on pt{111}
  • It may be used in the preparation of dimethyl 2,3-dimethylenebutanedioate

Specifications

CAS 600-22-6
Density 1.13 g/mL (at 25°C (literature))
Boiling Point 134°C to 137°C (lit.)
Molecular Formula C4H6O3
Refractive Index n20/D 1.404 (literature)
Linear Formula CH3COCO2CH3
MDL Number MFCD00008754
Quantity 100 g
Synonym Pyruvic acid methyl ester
Molecular Weight (g/mol) 102.09
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.