Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl quinoline-2-carboxylate | 19575-07-6 | MFCD00160630 | 99.4% | 187.19 g/mol | C11H9NO2 | 5 G
SDP

Catalog No. 5000302260
Encompass_Preferred

Methyl quinaldate (methyl quinoline-2-carboxylate) is a research-grade methyl ester of quinoline-2-carboxylic acid supplied as a solid. It is used for chemical and biological research and has reported antibacterial activity against Mycobacterium tuberculosis H37Rv (MIC 2 μg/mL).

  • Chemical name: methyl quinoline-2-carboxylate.
  • Molecular formula: C11H9NO2.
  • Molecular weight: 187.19 g/mol.
  • Purity: 99.4%.
  • Physical appearance: solid (powder).
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Intended use: research and screening applications (not for human use).
  • Biological activity: reported antibacterial activity vs Mycobacterium tuberculosis (MIC 2 μg/mL).

Catalog No. 50-003-02260 Supplier Medchemexpress LLC Supplier No. HYW0161415G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl quinaldate (methyl quinoline-2-carboxylate) is a research-grade methyl ester of quinoline-2-carboxylic acid supplied as a solid. It is used for chemical and biological research and has reported antibacterial activity against Mycobacterium tuberculosis H37Rv (MIC 2 μg/mL).

  • Chemical name: methyl quinoline-2-carboxylate.
  • Molecular formula: C11H9NO2.
  • Molecular weight: 187.19 g/mol.
  • Purity: 99.4%.
  • Physical appearance: solid (powder).
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Intended use: research and screening applications (not for human use).
  • Biological activity: reported antibacterial activity vs Mycobacterium tuberculosis (MIC 2 μg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.