Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate, 98%, Thermo Scientific™

Catalog No. AAH3123606
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3123606 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3123606 Supplier Thermo Scientific Chemicals Supplier No. H3123606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64700-65-8
Density 1.226
Boiling Point 83°C (0.25mmHg)
Molecular Formula C6H9NO3
Quantity 5g
MDL Number MFCD01318621
Sensitivity Moisture sensitive
Synonym r-methyl 5-oxopyrrolidine-2-carboxylate, methyl 2r-5-oxopyrrolidine-2-carboxylate, methyl r-2-pyrrolidone-5-carboxylate, h-d-pyr-ome, methyl r-pyroglutamate, methyl d-pyroglutamate, d-proline, 5-oxo-, methyl ester, methyl r---2-pyrrolidinone-5-carboxylate, d-pyroglutamic acid methyl ester, d-pyr-ome
InChI Key HQGPKMSGXAUKHT-SCSAIBSYSA-N
SMILES COC(=O)C1CCC(=O)N1
IUPAC Name methyl (2R)-5-oxopyrrolidine-2-carboxylate
Molecular Weight (g/mol) 143.142
PubChem CID 385461
Formula Weight 143.14
Percent Purity 98%
Chemical Name or Material Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate
Show More Show Less
TSCA No
Recommended Storage Keep cold; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.