Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methyl Red, 0.1% w/v solution in ethanol

Catalog No. AA38698AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38698AP 500 mL
AA38698AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38698AP Supplier Thermo Scientific Chemicals Supplier No. 038698AP
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 74886-84-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74886-84-3
MDL Number MFCD00002425

Specifications

Concentration 0.1% w/v solution in ethanol
UN Number UN1170
Quantity 500 mL
Physical Form Liquid
Formula Weight 269.3
Chemical Name or Material Methyl Red
Hazard Category H225-H302-H319-H331-H370
Hazard Statement GHS H Statement
H224
Extremely flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P311-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.