Learn More
Medchemexpress LLC Methyl red | 493-52-7 | MFCD00002425 | 98.7% | 269.30 g/mol | C15H15N3O2 | 100g
SDP
Catalog No. 5000269859
Encompass_Preferred
Methyl red is a pH-sensitive acid-base indicator and colorimetric reagent with a pKa of 5 1 Methyl red achieves visual judgment of the titration endpoint through reversible structural changes of protonation (red pH 4 4) and deprotonation (yellow pH 6 2) Methyl red is widely used in acid-base titration in chemical analysis and buffer pH monitoring in biochemistry[1][2]
Catalog No. 50-002-69859 Supplier Medchemexpress LLC Supplier No. HYY0699100G
Methyl red is a pH-sensitive acid-base indicator and colorimetric reagent with a pKa of 5 1 Methyl red achieves visual judgment of the titration endpoint through reversible structural changes of protonation (red pH 4 4) and deprotonation (yellow pH 6 2) Methyl red is widely used in acid-base titration in chemical analysis and buffer pH monitoring in biochemistry[1][2]