Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red Indicator, 0.02% Aqueous, Reagents

Catalog No. p-200022022
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-549 125 mL
01-336-548 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-549 Supplier Reagents Holdings Llc Supplier No. C7118600125C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl Red Indicator, 0.02% Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methyl Red Indicator, 0.02% Aqueous
Boiling Point 100°C
CAS 7732-18-5, 845-10-3
Packaging Glass Bottle
Specific Gravity 1.001
Synonym Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid
Color Orange to Red
Quantity 125 mL
Physical Form Liquid
Percent Purity 0.02%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.