Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red Indicator, 0.02% in Methanol, Reagents
Specifications
Specifications
| Chemical Name or Material | Methyl Red Indicator, 0.02% in Methanol |
| CAS | 67-56-1, 493-52-7 |
| Boiling Point | 65°C |
| Packaging | Poly Bottle |
| Specific Gravity | 0.7866 |
| Color | Red |
| Synonym | Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Percent Purity | 0.02% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?