Learn More
Methyl Red Indicator, 1% w/v Aqueous, Reagents
Specifications
Specifications
| Chemical Name or Material | Methyl Red Indicator, 1% w/v Aqueous |
| CAS | 7732-18-5, 845-10-3 |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.005 |
| Synonym | Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid |
| Color | Orange to Red |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Percent Purity | 1% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.