Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Methyl Red Indicator
SCI_ED

Catalog No. S25434A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25434A 25 g Solid
S25434 10 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25434A
Add to Cart
Add to Cart
This item is not returnable. View return policy
pH 4.8-6.0 (red-yellow)

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym Sodium Salt, Methyl Red In Alcohol
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Linear Formula C15H14N3O2Na
Formula Weight 291.28
Packaging Solid
Density 0.78g/mL
Color Orange
Quantity 25 g
Chemical Name or Material Methyl Red Indicator
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.