Learn More
Aqua Solutions Methyl Red 0.1% in Ethanol (pH 4.2 pink 6.2 yellow)(500ML)
SDP
Catalog No. NC9686002
Encompass_Preferred
Methyl Red 0.1% in Ethanol (pH 4.2 pink 6.2 yellow)(500ML)
Catalog No. NC9686002 Supplier Aqua Solutions Supplier No. 5851500ML
Methyl Red 0.1% in Ethanol (pH 4.2 pink 6.2 yellow)(500ML)