Learn More
Alkali Scientific Methyl Red Indicator Solution, 25 mL (1 oz), Amber Bottle, 1/Pk
SDP
Catalog No. 5000663168
Encompass_Preferred
Methyl Red Indicator Solution, 25 mL (1 oz), Amber Bottle, 1/Pk. The Methyl Red Indicator Solution is used for detecting mixed acid fermentation in bacterial cultures. It provides clear results for identifying organisms based on their ability to produce acidic byproducts during fermentation. The amber bottle ensures protection from light degradation, ensuring the solution remains effective for laboratory use.
Catalog No. 50-006-63168 Supplier Alkali Scientific Supplier No. MRIS00
Methyl Red Indicator Solution, 25 mL (1 oz), Amber Bottle, 1/Pk. The Methyl Red Indicator Solution is used for detecting mixed acid fermentation in bacterial cultures. It provides clear results for identifying organisms based on their ability to produce acidic byproducts during fermentation. The amber bottle ensures protection from light degradation, ensuring the solution remains effective for laboratory use.