Learn More
Methyl Red-Methylene Blue, Mixed Indicator, for Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Chemical Name or Material | Methyl Red-Methylene Blue |
| Melting Point | -115°C |
| Name Note | Mixed Indicator, for Nitrogen (Ammonia), Nitrogen (Organic) |
| Boiling Point | 78°C |
| CAS Max % | 89.88, 4.96, 4.96, 0.13, 0.07 |
| CAS | 67-63-0, 67-56-1, 845-10-3, 61-73-4 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Linear Formula | C15H14N3O2Na |
| Decomposition Information | Carbon monoxide; Carbon dioxide; May release flammable gases |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.