Learn More
Abcam Methyl red, pH indicator, 25G
Catalog No. 502704679
Encompass_Preferred
MW 269.3 g/mol. pH indicator. Azo dye used in the methyl red (MR) test to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose. The transition range is pH 4.4 (red) to pH 6.2 (yellow). Absorption: λmax 410 nm.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4679 Supplier Abcam Supplier No. AB14636825G
MW 269.3 g/mol. pH indicator. Azo dye used in the methyl red (MR) test to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose. The transition range is pH 4.4 (red) to pH 6.2 (yellow). Absorption: λmax 410 nm.
The product is subject to the following: Abcam Restricted Use Statement