Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red Sodium Salt, ACS Grade, LabChem™

Catalog No. LC170807 Shop All LabChem Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC170807 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC170807 Supplier LabChem, Inc. Supplier No. LC170807
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

CAS Max % 100
Identification Passes Test
Packaging Amber Glass
Linear Formula C15H14N3O2Na
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Brown/Red
Solubility Information Soluble in water
Quantity 10 g
Physical Form Powder
Formula Weight 291.28
Grade ACS
Chemical Name or Material Methyl Red Sodium Salt
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.