Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red Sodium Salt, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCM090010 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCM090010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCM090010 Supplier Ricca Chemical Company Supplier No. RDCM090010F1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Red to Violet
Quantity 10 g
Grade ACS
Chemical Name or Material Methyl Red Sodium Salt
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.