Learn More
Medchemexpress LLC Methyl ricinoleate | 141-24-2 | 25 MG
SDP
Catalog No. 5000420571
Encompass_Preferred
Methyl ricinoleate is a fatty acid methyl ester derived from ricinoleic acid, a monounsaturated omega-9 fatty acid found in castor oil. This biochemical reagent is suitable for life science-related research as a biological material or organic compound. It is frequently utilized as a reference for the analysis of fatty acid methyl esters via gas chromatography and has been explored for potential applications as a biofuel and a component in biodegradable plastics and surfactants.
  • Derived from ricinoleic acid, a monounsaturated omega-9 fatty acid.
  • Used as a reference for fatty acid methyl ester analysis by gas chromatography.
  • Potential application as a biofuel.
  • Component in biodegradable plastics and surfactants.
  • Biochemical reagent for life science research.
  • Colorless to light yellow liquid.
Catalog No. 50-004-20571 Supplier Medchemexpress LLC Supplier No. 25MGHYN8045
Add to Cart
Add to Cart
Methyl ricinoleate is a fatty acid methyl ester derived from ricinoleic acid, a monounsaturated omega-9 fatty acid found in castor oil. This biochemical reagent is suitable for life science-related research as a biological material or organic compound. It is frequently utilized as a reference for the analysis of fatty acid methyl esters via gas chromatography and has been explored for potential applications as a biofuel and a component in biodegradable plastics and surfactants.
  • Derived from ricinoleic acid, a monounsaturated omega-9 fatty acid.
  • Used as a reference for fatty acid methyl ester analysis by gas chromatography.
  • Potential application as a biofuel.
  • Component in biodegradable plastics and surfactants.
  • Biochemical reagent for life science research.
  • Colorless to light yellow liquid.