Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl ricinoleate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013070 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3070 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3070 Supplier MilliporeSigma Supelco Supplier No. 83916100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl ricinoleate is an enantiomerically renewable compound commercially obtained by the transesterification from castor oil, commonly used in cosmetics, plasticizers, lubricating oils and fine-chemical industries.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 141-24-2
Shipping Condition Ambient
Physical Form Neat
Linear Formula C19H36O3
MDL Number MFCD00046712
Synonym (R)-12-Hydroxy-cis-9-octadecenoic acid methyl ester; Methyl 12-hydroxyoleate; Ricinoleic acid methyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 312.49
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.