Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl rosmarinate | 99353-00-1 | 1 ML
SDP

Catalog No. 5000420007
Encompass_Preferred

Methyl rosmarinate is a noncompetitive tyrosinase inhibitor isolated from *Rabdosia serra*, with an IC50 of 0.28 mM for mushroom tyrosinase. It also inhibits α-glucosidase and is intended for research use only.

  • Noncompetitive tyrosinase inhibitor
  • Inhibits α-glucosidase
  • Isolated from *Rabdosia serra*
  • Molecular formula: C19H18O8
  • Molecular weight: 374.34
  • Appearance: Solid
  • Recommended storage: 4°C, protected from light
  • In solvent storage: -80°C for 6 months; -20°C for 1 month, protected from light
  • Shipping: Room temperature if less than 2 weeks
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation

Catalog No. 50-004-20007 Supplier Medchemexpress LLC Supplier No. HYN326610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl rosmarinate is a noncompetitive tyrosinase inhibitor isolated from *Rabdosia serra*, with an IC50 of 0.28 mM for mushroom tyrosinase. It also inhibits α-glucosidase and is intended for research use only.

  • Noncompetitive tyrosinase inhibitor
  • Inhibits α-glucosidase
  • Isolated from *Rabdosia serra*
  • Molecular formula: C19H18O8
  • Molecular weight: 374.34
  • Appearance: Solid
  • Recommended storage: 4°C, protected from light
  • In solvent storage: -80°C for 6 months; -20°C for 1 month, protected from light
  • Shipping: Room temperature if less than 2 weeks
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.