Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(-)-2-Isocyanato-3-methylbutyrate 98.0+%, TCI America™
SDP

Catalog No. I04661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04661G 1 g
I04665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04661G Supplier TCI America Supplier No. I04661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30293-86-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.169
MDL Number MFCD00191531
InChI Key JCVZXXSBCISMLJ-LURJTMIESA-N
Synonym (S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester, Methyl (S)-(-)-2-Isocyanatoisovalerate
PubChem CID 10397066
IUPAC Name methyl (2S)-2-isocyanato-3-methylbutanoate
SMILES CC(C)C(C(=O)OC)N=C=O

Specifications

Color Yellow
Boiling Point 76°C
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-2-Isocyanato-3-methylbutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.