Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl (S)-(-)-N-Z-Aziridine-2-Carboxylate

Catalog No. 5160315G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-603-15G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-603-15G Supplier Sigma Aldrich Supplier No. 5160315G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl (S)-(-)-N-Z-Aziridine-2-Carboxylate

Specifications

CAS 556-81-0
Molecular Formula C12H32N2O2
Refractive Index n20/D 1.519
Linear Formula (CH3)3N(OH)(CH2)6N(OH)(CH3)3
MDL Number MFCD00677130
Quantity 100 mL
Synonym N,N,N,N',N',N'-Hexamethylhexamethylenediammonium dihydroxide
Molecular Weight (g/mol) 236.39 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.