Learn More
Medchemexpress LLC Methyl salicylate | 119-36-8 | MFCD00002214 | 100.0% | 152.15 g/mol | C8H8O3 | 100g
SDP
Catalog No. 5000268582
Encompass_Preferred
Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent Also used as a pesticide a denaturant a fragrance ingredient and a flavoring agent in food and tobacco products[1] A systemic acquired resistance (SAR) signal in tobacco[2] A topical nonsteroidal anti-inflammatory agent (NSAID) Methyl salicylate lactoside is a COX inhibitor[4]
Catalog No. 50-002-68582 Supplier Medchemexpress LLC Supplier No. HYY0189100G
Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent Also used as a pesticide a denaturant a fragrance ingredient and a flavoring agent in food and tobacco products[1] A systemic acquired resistance (SAR) signal in tobacco[2] A topical nonsteroidal anti-inflammatory agent (NSAID) Methyl salicylate lactoside is a COX inhibitor[4]