Learn More
Selleck Chemical LLC Methyl salicylate S3756-5mg
SDP
Catalog No. 503108952
Encompass_Preferred
Methyl salicylate (Betula oil Gaultheria oil Methyl 2-hydroxybenzoate Natural wintergreen oil) is an organic ester naturally produced by many species of plants particularly wintergreens It is an agonist for TRPA1 and TRPV1
Catalog No. 50-310-8952 Supplier Selleck Chemical LLC Supplier No. S37565MG
Methyl salicylate (Betula oil Gaultheria oil Methyl 2-hydroxybenzoate Natural wintergreen oil) is an organic ester naturally produced by many species of plants particularly wintergreens It is an agonist for TRPA1 and TRPV1