Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl salicylate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012721 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2721 1 mL
11-101-2722 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2721 Supplier MilliporeSigma Supelco Supplier No. 766311MLF
Add to Cart
Add to Cart

Methyl Salicylate is a methyl ester of salicylic acid, widely used as phytohormones and defensive signaling compounds. It is most commonly used in topical products as a rubefacients.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 847°F
CAS 119-36-8
Vapor Pressure 1 mmHg (54°C)
Vapor Density 5.26 (vs. air)
Density 1.174 g/mL (at 25°C)
Flash Point 96°C - Closed Cup
Physical Form Neat
Linear Formula 2-(HO)C6H4 CO2 CH3
MDL Number MFCD00002214
Melting Point -8°C to -7°C
Boiling Point 222°C
Synonym 2-Hydroxybenzoic acid methyl ester; Methyl 2-hydroxybenzoate; Oil of wintergreen; Wintergreen oil
Type Analytical Standard
Quantity 1 mL
Formula Weight 152.15
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.