Learn More
Cayman Chemical Methyl Salicylate-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1219802-12-6, 10 mg
SDP
Catalog No. 502943387
Encompass_Preferred
An internal standard for the quantification of methyl salicylate by GC- or LC-MS
Catalog No. 50-294-3387 Supplier Cayman Chemical Supplier No. 4214510 MG
An internal standard for the quantification of methyl salicylate by GC- or LC-MS