Learn More
Medchemexpress LLC Methyl salicylate-d4 | 1219802-12-6 | 156.17 | C8H4D4O3 | 25mg
SDP
Catalog No. 5000269149
Encompass_Preferred
Methyl Salicylate-d4 is the deuterium labeled Methyl Salicylate[1] Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent Also used as a pesticide a denaturant a fragrance ingredient and a flavoring agent in food and tobacco products[2] A systemic acquired resistance (SAR) signal in tobacco[3] A topical nonsteroidal anti-inflammatory agent (NSAID) Methyl salicylate lactoside is a COX inhibitor[5]
Catalog No. 50-002-69149 Supplier Medchemexpress LLC Supplier No. HYY0189S25MG
Add to Cart
Add to Cart
Methyl Salicylate-d4 is the deuterium labeled Methyl Salicylate[1] Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent Also used as a pesticide a denaturant a fragrance ingredient and a flavoring agent in food and tobacco products[2] A systemic acquired resistance (SAR) signal in tobacco[3] A topical nonsteroidal anti-inflammatory agent (NSAID) Methyl salicylate lactoside is a COX inhibitor[5]