Learn More
Medchemexpress LLC Methyl salicylate (salicylic acid methyl ester) | 119-36-8 | MFCD00002214 | 100.0% | 152.15 g/mol | C8H8O3 | 500mg
SDP
Catalog No. 5000269148
Encompass_Preferred
Methyl Salicylate (Standard) is the analytical standard of Methyl Salicylate This product is intended for research and analytical applications Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent Also used as a pesticide a denaturant a fragrance ingredient and a flavoring agent in food and tobacco products[1] A systemic acquired resistance (SAR) signal in tobacco[2] A topical nonsteroidal anti-inflammatory agent (NSAID) Methyl salicylate lactoside is a COX inhibitor[4]
Catalog No. 50-002-69148 Supplier Medchemexpress LLC Supplier No. HYY0189R500MG
Methyl Salicylate (Standard) is the analytical standard of Methyl Salicylate This product is intended for research and analytical applications Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent Also used as a pesticide a denaturant a fragrance ingredient and a flavoring agent in food and tobacco products[1] A systemic acquired resistance (SAR) signal in tobacco[2] A topical nonsteroidal anti-inflammatory agent (NSAID) Methyl salicylate lactoside is a COX inhibitor[4]