Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl sinapate | 20733-94-2 | 10 MM 1 ML
SDP

Catalog No. 5000420503
Encompass_Preferred

Methyl sinapate (MSA) is a hydroxycinnamic acid and a natural UV screening agent, typically appearing as a pale purple to purple solid. This compound is suitable for various research applications requiring a pure, characterized reagent.

  • Molecular formula: C12H14O5
  • Molecular weight: 238.24
  • Minimum purity: 95.0%
  • Derived from microorganisms, specifically *Fusarium oxysporum*
  • Available as a 10 mM 1 mL solution in DMSO

Catalog No. 50-004-20503 Supplier Medchemexpress LLC Supplier No. HYN326510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl sinapate (MSA) is a hydroxycinnamic acid and a natural UV screening agent, typically appearing as a pale purple to purple solid. This compound is suitable for various research applications requiring a pure, characterized reagent.

  • Molecular formula: C12H14O5
  • Molecular weight: 238.24
  • Minimum purity: 95.0%
  • Derived from microorganisms, specifically *Fusarium oxysporum*
  • Available as a 10 mM 1 mL solution in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.