Learn More
Medchemexpress LLC Methyl stearate | 112-61-8 | 99.3% | 500 MG
SDP
Catalog No. 5000409667
Encompass_Preferred
Methyl stearate, isolated from Rheum palmatum L., is a component of soybean and rapeseed biodiesels. It inhibits the egg hatching of root-knot nematode (Meloidogyne incognita) and has a repellent effect on second-stage larvae (J2s), thereby reducing nematode infection of plants.
  • Component of soybean and rapeseed biodiesels
  • Inhibits egg hatching of root-knot nematode
  • Repellent effect on second-stage larvae (J2s)
  • Reduces nematode infection of plants
Catalog No. 50-004-09667 Supplier Medchemexpress LLC Supplier No. HYB1934500MG
Methyl stearate, isolated from Rheum palmatum L., is a component of soybean and rapeseed biodiesels. It inhibits the egg hatching of root-knot nematode (Meloidogyne incognita) and has a repellent effect on second-stage larvae (J2s), thereby reducing nematode infection of plants.
  • Component of soybean and rapeseed biodiesels
  • Inhibits egg hatching of root-knot nematode
  • Repellent effect on second-stage larvae (J2s)
  • Reduces nematode infection of plants