Learn More
Medchemexpress LLC Methyl stearate | 112-61-8 | MFCD00009005 | 99.31% | 1 G
SDP
Catalog No. 5000411090
Encompass_Preferred
Methyl stearate, also known as stearic acid methyl ester, is a compound isolated from *Rheum palmatum* L. and is a component of soybean and rapeseed biodiesels. It exhibits biological activity by inhibiting the egg hatching of root-knot nematode (*Meloidogyne incognita*) and has a repellent effect on second-stage larvae, thereby reducing nematode infection in plants.
  • Isolated from *Rheum palmatum* L.
  • Component of soybean and rapeseed biodiesels.
  • Inhibits egg hatching of root-knot nematode.
  • Repellent effect on second-stage larvae (J2s).
  • Reduces nematode infection of plants.
  • Soluble in DMSO (requires ultrasonication).
Catalog No. 50-004-11090 Supplier Medchemexpress LLC Supplier No. HYB19341G
Methyl stearate, also known as stearic acid methyl ester, is a compound isolated from *Rheum palmatum* L. and is a component of soybean and rapeseed biodiesels. It exhibits biological activity by inhibiting the egg hatching of root-knot nematode (*Meloidogyne incognita*) and has a repellent effect on second-stage larvae, thereby reducing nematode infection in plants.
  • Isolated from *Rheum palmatum* L.
  • Component of soybean and rapeseed biodiesels.
  • Inhibits egg hatching of root-knot nematode.
  • Repellent effect on second-stage larvae (J2s).
  • Reduces nematode infection of plants.
  • Soluble in DMSO (requires ultrasonication).