Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl stearate 96 fG5KG
Catalog No. 502901966
Encompass
Methyl stearate is one of the key fatty acid methyl ester (fAME) component of biodiesel fuel produced by the transesterification of triglyceride oil and a primary alcohol. It is reported to occur in the volatile oil of Rheum palmatum L rhizomes and Ziziphus spina-christi leaves.
Catalog No. 50-290-1966 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W5048075KG
Methyl stearate is one of the key fatty acid methyl ester (fAME) component of biodiesel fuel produced by the transesterification of triglyceride oil and a primary alcohol. It is reported to occur in the volatile oil of Rheum palmatum L rhizomes and Ziziphus spina-christi leaves.