Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl stearate 99 GC50G
Catalog No. 502902665
Encompass
Methyl stearate or methyl octadecanoic acid is a saturated methyl ester and a representative key molecule of biodiesel fatty acid methyl esters (fAMEs) produced from vegetable oils. Methyl stearate occurs naturally in clove oil.
Catalog No. 50-290-2665 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S537650G
Methyl stearate or methyl octadecanoic acid is a saturated methyl ester and a representative key molecule of biodiesel fatty acid methyl esters (fAMEs) produced from vegetable oils. Methyl stearate occurs naturally in clove oil.