Learn More
Medchemexpress LLC Methyl stearate-d35 | 29823-25-4 | 99.9% | 100 MG
SDP
Catalog No. 5000411087
Encompass_Preferred
Methyl stearate-d35 is the deuterium labeled Methyl stearate. Methyl stearate, isolated from Rheum palmatum L., is a component of soybean and rapeseed biodiesels. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11087 Supplier Medchemexpress LLC Supplier No. HYB1934S1100MG
Methyl stearate-d35 is the deuterium labeled Methyl stearate. Methyl stearate, isolated from Rheum palmatum L., is a component of soybean and rapeseed biodiesels. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.