Learn More
Medchemexpress LLC Methyl stearate-d35 | 29823-25-4 | 99.9% | 50 MG
SDP
Catalog No. 5000411190
Encompass_Preferred
Methyl stearate-d35 is the deuterium-labeled version of Methyl stearate (HY-B1934), a compound isolated from Rheum palmatum L. and a component of soybean and rapeseed biodiesels. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for use as tracers in drug development, and deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11190 Supplier Medchemexpress LLC Supplier No. HYB1934S150MG
Methyl stearate-d35 is the deuterium-labeled version of Methyl stearate (HY-B1934), a compound isolated from Rheum palmatum L. and a component of soybean and rapeseed biodiesels. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for use as tracers in drug development, and deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.