Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl stearate, mixture of homologs

Catalog No. p-168579
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304220010 1 kg Glass Bottle
AC304220025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC304220010 Supplier Thermo Scientific Chemicals Supplier No. 304220010
Only null left
Add to Cart

CAS: 112-61-8 | C19H38O2 | 298.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-61-8
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00009005
InChI Key HPEUJPJOZXNMSJ-UHFFFAOYSA-N
Synonym methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester
PubChem CID 8201
ChEBI CHEBI:69188
IUPAC Name methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Melting Point 37.0°C to 39.0°C
Color Cream to White or Yellow
Density 0.8360g/mL
Boiling Point 215.0°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Linear Formula CH3(CH2)16CO2CH3
Quantity 1 kg
Saponification Value 186 to 192mg KOH/g
Specific Gravity 0.836
Merck Index 15,893
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 298.51
Physical Form Paste
Chemical Name or Material Methyl stearate
Show More Show Less
EINECSNumber 203-990-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.