Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Succinamate 98.0+%, TCI America™
SDP

Catalog No. S047925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S047925G 25 g
S04795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S047925G Supplier TCI America Supplier No. S047925G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53171-39-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
InChI Key HURZMSZDVGMYKJ-UHFFFAOYSA-N
Synonym Succinamic Acid Methyl Ester
PubChem CID 12550576
IUPAC Name methyl 4-amino-4-oxobutanoate
SMILES COC(=O)CCC(=O)N

Specifications

Melting Point 89°C
Color White
Boiling Point 118°C
Quantity 25 g
Formula Weight 131.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Succinamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.